Assay Details
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145188.1 | NP_001138660.1 | 1 - 2 | 102 | 113 | |
GenBank mRNA | FJ546078.1 | - | 1 - 2 | 102 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
