Assay Details
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162553.2 | NP_001156025.1 | 2 - 3 | 164 | 123 | |
GenBank mRNA | EE773754.1 | - | 1 - 2 | 152 | 123 | |
EE834734.1 | - | 2 - 3 | 197 | 123 | ||
FJ969414.1 | - | 1 - 2 | 145 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
