Assay Details
Gene Symbol: ADCYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009776.1 | NP_001009776.1 | 3 - 4 | 812 | 78 | |
GenBank mRNA | S83511.1 | - | 3 - 4 | 812 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
