Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009806.1 | NP_001009806.1 | 1 - 2 | 145 | 96 | |
GenBank mRNA | X53934.1 | - | 145 | 96 | ||
X55641.1 | - | 145 | 96 | |||
X60148.1 | - | 85 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
