Assay Details
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038016.1 | NP_001033105.1 | 2 - 3 | 400 | 93 | |
XM_012174287.3 | XP_012029677.1 | 3 - 4 | 551 | 93 | ||
XM_012174398.3 | XP_012029788.1 | 3 - 4 | 530 | 93 | ||
XM_012174640.3 | XP_012030030.1 | 2 - 3 | 389 | 93 | ||
XM_027969192.1 | XP_027824993.1 | 3 - 4 | 551 | 93 | ||
XM_027969222.1 | XP_027825023.1 | 3 - 4 | 551 | 93 | ||
XM_027969253.1 | XP_027825054.1 | 3 - 4 | 530 | 93 | ||
XM_027969342.1 | XP_027825143.1 | 3 - 4 | 551 | 93 | ||
XM_027969414.1 | XP_027825215.1 | 3 - 4 | 551 | 93 | ||
XM_027969464.1 | XP_027825265.1 | 3 - 4 | 530 | 93 | ||
XM_027969518.1 | XP_027825319.1 | 3 - 4 | 551 | 93 | ||
XM_027969576.1 | XP_027825377.1 | 3 - 4 | 551 | 93 | ||
XM_027969620.1 | XP_027825421.1 | 3 - 4 | 530 | 93 | ||
GenBank mRNA | AY395982.3 | - | 2 - 3 | 400 | 93 | |
AY395983.3 | - | 2 - 3 | 400 | 93 | ||
AY445823.3 | - | 2 - 3 | 400 | 93 | ||
AY445824.3 | - | 2 - 3 | 400 | 93 | ||
AY491977.4 | - | 2 - 3 | 421 | 93 | ||
DQ304075.1 | - | 2 - 3 | 406 | 93 | ||
DQ304076.1 | - | 2 - 3 | 400 | 93 | ||
DQ304077.1 | - | 2 - 3 | 421 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
