Assay Details
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141954.2 | NP_001135426.1 | 6 - 7 | 454 | 105 | |
GenBank mRNA | FJ374172.1 | - | 6 - 7 | 456 | 105 | |
KJ710702.1 | - | 6 - 7 | 454 | 105 | ||
KJ710706.1 | - | 6 - 7 | 454 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
