Assay Details
Gene Symbol: PRDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280704.1 | NP_001267633.1 | 2 - 3 | 342 | 68 | |
GenBank mRNA | KC342239.1 | - | 2 - 3 | 342 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
