Assay Details
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075101.1 | NP_001068569.1 | 3 - 4 | 340 | 119 | |
GenBank mRNA | DQ054533.1 | - | 3 - 4 | 340 | 119 |

More Information
Set Membership: |
![]() ![]() |
