Assay Details
Gene Symbol: YBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093786.1 | NP_001087255.1 | 2 | 772 | 85 | |
XM_015091592.2 | XP_014947078.2 | 6 - 7 | 880 | 85 | ||
XM_015091593.2 | XP_014947079.2 | 5 - 6 | 784 | 85 | ||
GenBank mRNA | EF488163.1 | - | 2 | 772 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
