Assay Details
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139446.2 | NP_001132918.1 | 5 - 6 | 680 | 160 | |
GenBank mRNA | FJ223570.1 | - | 5 - 6 | 681 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() |
