Assay Details
Gene Symbol: HNRNPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971131.1 | XP_027826932.1 | 3 - 4 | 757 | 97 | |
XM_027971132.1 | XP_027826933.1 | 2 - 3 | 700 | 97 | ||
XM_027971133.1 | XP_027826934.1 | 3 - 4 | 756 | 97 | ||
XM_027971134.1 | XP_027826935.1 | 2 - 3 | 699 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
