Assay Details
Gene Symbol: DOP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097311.2 | XP_014952797.2 | 36 - 37 | 7062 | 94 | |
XM_027972311.1 | XP_027828112.1 | 36 - 37 | 7062 | 94 | ||
XM_027972312.1 | XP_027828113.1 | 35 - 36 | 7032 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
