Assay Details
Gene Symbol: LOC101117028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091984.2 | XP_014947470.2 | 8 - 9 | 942 | 66 | |
XM_027970323.1 | XP_027826124.1 | 8 - 9 | 1452 | 66 | ||
XM_027970329.1 | XP_027826130.1 | 7 - 8 | 849 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
