Assay Details
Gene Symbol: LOC101117028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091984.2 | XP_014947470.2 | 9 - 10 | 989 | 71 | |
XM_027970323.1 | XP_027826124.1 | 9 - 10 | 1499 | 71 | ||
XM_027970329.1 | XP_027826130.1 | 8 - 9 | 896 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
