Assay Details
Gene Symbol: LOC101103602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001935.4 | XP_004001984.1 | 12 - 13 | 1457 | 102 | |
XM_012130794.3 | XP_011986184.1 | 12 - 13 | 1457 | 102 | ||
XM_012131126.3 | XP_011986516.1 | 8 - 9 | 943 | 102 | ||
XM_012131595.3 | XP_011986985.1 | 7 - 8 | 789 | 102 | ||
XM_027969579.1 | XP_027825380.1 | 12 - 13 | 1494 | 102 | ||
XM_027969591.1 | XP_027825392.1 | 12 - 13 | 1493 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
