Assay Details
Gene Symbol: LOC101103602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001935.4 | XP_004001984.1 | 1 - 2 | 281 | 70 | |
XM_012130794.3 | XP_011986184.1 | 1 - 2 | 281 | 70 | ||
XM_012131126.3 | XP_011986516.1 | 1 - 2 | 274 | 70 | ||
XM_012131207.3 | XP_011986597.1 | 1 - 2 | 305 | 70 | ||
XM_012131519.3 | XP_011986909.1 | 1 - 2 | 324 | 70 | ||
XM_012131595.3 | XP_011986985.1 | 1 - 2 | 270 | 70 | ||
XM_027969579.1 | XP_027825380.1 | 1 - 2 | 318 | 70 | ||
XM_027969585.1 | XP_027825386.1 | 1 - 2 | 326 | 70 | ||
XM_027969591.1 | XP_027825392.1 | 1 - 2 | 317 | 70 | ||
XM_027969596.1 | XP_027825397.1 | 1 - 2 | 325 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
