Assay Details
Gene Symbol: LOC101103602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001935.4 | XP_004001984.1 | 3 - 4 | 537 | 60 | |
XM_012130794.3 | XP_011986184.1 | 3 - 4 | 537 | 60 | ||
XM_012131207.3 | XP_011986597.1 | 3 - 4 | 561 | 60 | ||
XM_012131519.3 | XP_011986909.1 | 3 - 4 | 580 | 60 | ||
XM_027969579.1 | XP_027825380.1 | 3 - 4 | 574 | 60 | ||
XM_027969585.1 | XP_027825386.1 | 3 - 4 | 582 | 60 | ||
XM_027969591.1 | XP_027825392.1 | 3 - 4 | 573 | 60 | ||
XM_027969596.1 | XP_027825397.1 | 3 - 4 | 581 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
