Assay Details
Gene Symbol: FGF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963901.1 | XP_027819702.1 | 4 - 5 | 355 | 85 | |
XM_027963902.1 | XP_027819703.1 | 4 - 5 | 322 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
