Assay Details
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098600.2 | XP_014954086.1 | 2 - 3 | 808 | 87 | |
XM_027975073.1 | XP_027830874.1 | 3 - 4 | 616 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
