Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972398.1 | XP_027828199.1 | 6 - 7 | 4439 | 79 | |
XM_027972399.1 | XP_027828200.1 | 5 - 6 | 4351 | 79 | ||
XM_027972400.1 | XP_027828201.1 | 5 - 6 | 4152 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
