Assay Details
Gene Symbol: RIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978085.1 | XP_027833886.1 | 21 - 22 | 4081 | 73 | |
XM_027978086.1 | XP_027833887.1 | 21 - 22 | 4078 | 73 | ||
XM_027978087.1 | XP_027833888.1 | 21 - 22 | 4057 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
