Assay Details
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959040.1 | XP_027814841.1 | 2 - 3 | 1257 | 92 | |
XM_027959041.1 | XP_027814842.1 | 2 - 3 | 498 | 92 | ||
XM_027959042.1 | XP_027814843.1 | 2 - 3 | 810 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
