Assay Details
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009924.4 | XP_004009973.2 | 2 - 3 | 269 | 73 | |
XM_027971094.1 | XP_027826895.1 | 2 - 3 | 220 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
