Assay Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956265.1 | XP_027812066.1 | 5 - 6 | 1920 | 97 | |
GenBank mRNA | JN598905.1 | - | 2 - 3 | 353 | 97 |
Gene Symbol: LOC114116628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973704.1 | XP_027829505.1 | 1 | 611 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
