Assay Details
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974421.1 | XP_027830222.1 | 8 - 9 | 1189 | 56 | |
XM_027974422.1 | XP_027830223.1 | 10 - 11 | 1255 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
