Assay Details
Gene Symbol: LOC101102548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012142060.3 | XP_011997450.2 | 34 - 35 | 4164 | 66 | |
XM_012142071.3 | XP_011997461.2 | 34 - 35 | 4164 | 66 | ||
XM_012142079.3 | XP_011997469.2 | 34 - 35 | 4164 | 66 | ||
XM_027965072.1 | XP_027820873.1 | 34 - 35 | 4164 | 66 | ||
XM_027965074.1 | XP_027820875.1 | 33 - 34 | 4071 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
