Assay Details
Gene Symbol: MACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007779.4 | XP_004007828.1 | 4 - 5 | 2504 | 106 | |
XM_012176466.3 | XP_012031856.1 | 5 - 6 | 2605 | 106 | ||
XM_027968502.1 | XP_027824303.1 | 4 - 5 | 3229 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
