Assay Details
Gene Symbol: ILRUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102663.2 | XP_014958149.1 | 2 - 3 | 510 | 102 | |
XM_015102664.2 | XP_014958150.1 | 2 - 3 | 510 | 102 | ||
XM_015102667.2 | XP_014958153.1 | 2 - 3 | 491 | 102 |

More Information
Set Membership: |
![]() ![]() |
