Assay Details
Gene Symbol: ARL6IP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004691.4 | XP_004004740.1 | 2 - 3 | 514 | 125 | |
XM_027964953.1 | XP_027820754.1 | 2 - 3 | 1168 | 125 | ||
XM_027964954.1 | XP_027820755.1 | 2 - 3 | 520 | 125 | ||
XM_027964955.1 | XP_027820756.1 | 2 - 3 | 520 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
