Assay Details
Gene Symbol: AIFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012105214.3 | XP_011960604.2 | 8 - 9 | 1075 | 60 | |
XM_012105216.3 | XP_011960606.2 | 8 - 9 | 1059 | 60 | ||
XM_012105217.3 | XP_011960607.2 | 8 - 9 | 1093 | 60 | ||
XM_027962446.1 | XP_027818247.1 | 8 - 9 | 1084 | 60 | ||
XM_027962447.1 | XP_027818248.1 | 8 - 9 | 1025 | 60 | ||
XM_027962448.1 | XP_027818249.1 | 8 - 9 | 1035 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
