Assay Details
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975128.1 | XP_027830929.1 | 9 - 10 | 1589 | 66 | |
XM_027975129.1 | XP_027830930.1 | 9 - 10 | 1425 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
