Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961380.1 | XP_027817181.1 | 11 - 12 | 2372 | 71 | |
XM_027961381.1 | XP_027817182.1 | 11 - 12 | 2363 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
