Assay Details
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967005.1 | XP_027822806.1 | 7 - 8 | 3179 | 63 | |
XM_027967006.1 | XP_027822807.1 | 7 - 8 | 3179 | 63 | ||
XM_027967007.1 | XP_027822808.1 | 7 - 8 | 3179 | 63 | ||
XM_027967008.1 | XP_027822809.1 | 8 - 9 | 1421 | 63 | ||
XM_027967010.1 | XP_027822811.1 | 8 - 9 | 1181 | 63 | ||
XM_027967011.1 | XP_027822812.1 | 7 - 8 | 3179 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
