Assay Details
Gene Symbol: LOC101111528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965195.1 | XP_027820996.1 | 4 - 5 | 1305 | 74 | |
XM_027965196.1 | XP_027820997.1 | 4 - 5 | 1316 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
