Assay Details
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017743.4 | XP_004017792.1 | 4 - 5 | 553 | 60 |
Gene Symbol: LOC114112206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963748.1 | XP_027819549.1 | 1 - 2 | 1127 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
