Assay Details
Gene Symbol: HYAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018442.4 | XP_004018491.1 | 3 - 4 | 1132 | 118 | |
XM_012100028.3 | XP_011955418.1 | 3 - 4 | 329 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
