Assay Details
Gene Symbol: CYP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976458.1 | XP_027832259.1 | 7 - 8 | 992 | 101 | |
XM_027976459.1 | XP_027832260.1 | 7 - 8 | 992 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
