Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010571.3 | XP_004010620.2 | 7 - 8 | 858 | 69 | |
XM_012181465.3 | XP_012036855.2 | 7 - 8 | 834 | 69 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010571.3 | XP_004010620.2 | 7 - 8 | 858 | 69 | |
XM_012181465.3 | XP_012036855.2 | 7 - 8 | 834 | 69 |