Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010571.3 | XP_004010620.2 | 8 - 9 | 994 | 67 | |
XM_012181465.3 | XP_012036855.2 | 8 - 9 | 970 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
