Assay Details
Gene Symbol: LOC105607297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971828.1 | XP_027827629.1 | 6 - 7 | 2690 | 80 | |
XM_027971829.1 | XP_027827630.1 | 7 - 8 | 2463 | 80 | ||
XM_027971830.1 | XP_027827631.1 | 2 - 3 | 771 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
