Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005590.3 | XP_004005639.1 | 3 - 4 | 2377 | 88 | |
XM_012161685.2 | XP_012017075.1 | 4 - 5 | 2446 | 88 | ||
XM_012161700.2 | XP_012017090.1 | 2 - 3 | 2230 | 88 | ||
XM_012161705.3 | XP_012017095.1 | 2 - 3 | 2491 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
