Assay Details
Gene Symbol: SEPTIN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971109.1 | XP_027826910.1 | 7 - 8 | 1086 | 73 | |
XM_027971110.1 | XP_027826911.1 | 7 - 8 | 1086 | 73 | ||
XM_027971111.1 | XP_027826912.1 | 7 - 8 | 1086 | 73 | ||
XM_027971113.1 | XP_027826914.1 | 7 - 8 | 1086 | 73 | ||
XM_027971114.1 | XP_027826915.1 | 7 - 8 | 1086 | 73 | ||
XM_027971115.1 | XP_027826916.1 | 7 - 8 | 1086 | 73 | ||
XM_027971116.1 | XP_027826917.1 | 7 - 8 | 1086 | 73 | ||
XM_027971117.1 | XP_027826918.1 | 7 - 8 | 1086 | 73 | ||
XM_027971118.1 | XP_027826919.1 | 7 - 8 | 1086 | 73 | ||
XM_027971119.1 | XP_027826920.1 | 7 - 8 | 1086 | 73 | ||
XM_027971120.1 | XP_027826921.1 | 7 - 8 | 1086 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
