Assay Details
Gene Symbol: SNAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010251.4 | XP_004010300.1 | 1 - 2 | 860 | 91 | |
XM_012180801.3 | XP_012036191.1 | 1 - 2 | 871 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
