Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 6 - 7 | 1171 | 73 | |
XM_027975770.1 | XP_027831571.1 | 6 - 7 | 1173 | 73 | ||
XM_027975771.1 | XP_027831572.1 | 6 - 7 | 1172 | 73 | ||
XM_027975772.1 | XP_027831573.1 | 6 - 7 | 1169 | 73 | ||
XM_027975775.1 | XP_027831576.1 | 6 - 7 | 1329 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
