Assay Details
Gene Symbol: GRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956669.1 | XP_027812470.1 | 7 - 8 | 580 | 68 | |
XM_027956670.1 | XP_027812471.1 | 7 - 8 | 580 | 68 | ||
XM_027956671.1 | XP_027812472.1 | 5 - 6 | 357 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
