Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 2 - 3 | 276 | 97 | |
XM_027958477.1 | XP_027814278.1 | 1 - 2 | 248 | 97 | ||
XM_027958478.1 | XP_027814279.1 | 2 - 3 | 276 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
