Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 30 - 31 | 4827 | 97 | |
XM_012100634.2 | XP_011956024.1 | 26 - 27 | 4062 | 97 | ||
XM_027958477.1 | XP_027814278.1 | 29 - 30 | 4799 | 97 | ||
XM_027958478.1 | XP_027814279.1 | 31 - 32 | 4731 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
