Assay Details
Gene Symbol: LOC114111483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963255.1 | XP_027819056.1 | 1 | 587 | 75 |
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980039.1 | XP_027835840.1 | 4 - 5 | 735 | 75 | |
XM_027980045.1 | XP_027835846.1 | 4 - 5 | 643 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
