Assay Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092365.2 | XP_014947851.2 | 2 - 3 | 414 | 106 |

More Information
Set Membership: |
![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092365.2 | XP_014947851.2 | 2 - 3 | 414 | 106 |
Set Membership: |
![]() ![]() |