Assay Details
Gene Symbol: IQCA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968407.1 | XP_027824208.1 | 16 - 17 | 2114 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IQCA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968407.1 | XP_027824208.1 | 16 - 17 | 2114 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() |